Millicell® Cell Culture Inserts, Hanging, PET Membrane
Supplier: Merck
Certificates
About this item
With Millicell inserts, attachment or suspension cells can access media from both their apical and basolateral sides. Cell growth, structure, and function more closely mimic what occurs in vivo. In addition, Millicell inserts make it possible to study both sides of the cell monolayer.
- Hanging inserts ideal for co-culturing and permeability assays
The inserts are easily prepared for SEM and TEM visualising techniques, and they are compatible with cellular and/or fluorescent stains.
Specifications
- Catalog No:
- MILXPIRP12R48
- MILXPIRP30R48
- Effective diameter of membrane:
- 6,5 mm
- 24 mm