Order Entry
Netherlands
ContactUsLinkComponent
Ytterbium, sputtering target, Ø 50.8 mm, Thickness: 1.59 mm
Ytterbium, sputtering target, Ø 50.8 mm, Thickness: 1.59 mm
Catalog # 45620.KS
CAS Number:  
Ytterbium, sputtering target, Ø 50.8 mm, Thickness: 1.59 mm
Catalog # 45620.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :173.04 g/mol
Merck Index:12,10239
Boiling Pt:1427 °C (1013 hPa)
Melting Pt:824 °C
Density:6.977 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-173-2
CAS Number:7440-64-4
Formula:Yb
MDL Number:MFCD00011286