Order Entry
Netherlands
ContactUsLinkComponent
Iron(III) chloride hexahydrate
Iron(III) chloride hexahydrate
Catalog # HACH2171820
Supplier:  Hach
CAS Number:  
undefined
Iron(III) chloride hexahydrate
Catalog # HACH2171820
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    20
  • Seq:
    0020

Specifications

About this item

 :
 :270.3 g/mol
Merck Index:13,04048
Boiling Pt:280…285 °C (1013 hPa)
Melting Pt:37 °C
Density:1.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
 :GHS05
 :GHS07
CAS Number:10025-77-1
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712