



Ammonia 7 N in methanol
:7664-41-7
:
:NH₃
:MFCD00011418
:17.03 g/mol
:0.77 g/cm³
:14 °C
:Refrigerator
:215-647-6
:3286
:3,II
:GHS06
:GHS08
:GHS05
:GHS02
- Appearance:Clear colorless to light yellow Solution
- Water:≤0.5 %
- Strength:6.0 to 7.5 N
- NMR (31P-NMR):Consistent with structure
- Elemental Analysis:52.5 to 56.9 % C
Frequently Bought Together