Order Entry
Netherlands
ContactUsLinkComponent
Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished one side 100 orientation
Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished one side 100 orientation
Catalog # 38501.KU
CAS Number:  
Aluminium lanthanum (III) oxide substrate 10x10x0.5 mm, polished one side 100 orientation
Catalog # 38501.KU
CAS Number:  

Specifications

  • Pk:
    5
  • Seq:
    0005
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Lanthanum(III) aluminum oxide, Lanthanum Aluminium oxide, Aluminium lanthanum trioxide
 :
 :213.89 g/mol
Storage Temperature:Ambient
EINECS:234-433-3
CAS Number:12003-65-5
Formula:AlLaO₃
MDL Number:MFCD00144550