4-Aminohippuric acid, Sigma-Aldrich®
Catalog # 8.14621.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Solubility:(20 °C) slightly soluble
- Melting Point:199 - 202 °C (decomposition)
- Molar Mass:194.18 g/mol
- Bulk Density:380 kg/m3
- pH:3.0 - 3.5 (20 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:194.19 g/mol
Merck Index:13,00441
Boiling Pt:441 °C (1013 hPa)
Melting Pt:199…202 °C
Density:1.402 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-518-9
CAS Number:61-78-9
Formula:H₂NC₆H₄C(O)NHCH₂COOH
MDL Number:MFCD00007890