Order Entry
Netherlands
ContactUsLinkComponent
4-Aminohippuric acid Reag. Ph. Eur. 1003701
4-Aminohippuric acid Reag. Ph. Eur. 1003701
Catalog # 87973.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
4-Aminohippuric acid Reag. Ph. Eur. 1003701
Catalog # 87973.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :194.19 g/mol
EINECS:000-000-0
ADR:3,II
CAS Number:61-78-9
Formula:H₂NC₆H₄C(O)NHCH₂COOH
MDL Number:MFCD00007890