4-Aminohippuric acid 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61-78-9
:
Formula:H₂NC₆H₄C(O)NHCH₂COOH
MDL Number:MFCD00007890
:194.19 g/mol
Merck Index:13,00441
Boiling Pt:441 °C (1013 hPa)
Melting Pt:198…202 °C
Density:1,402 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-518-9
Specification Test Results
- Appearance:White to beige to grey to light brown Powder
- Infrared spectrum:Conforms
- Melting point:197 to 204 °C (Dec)
- Titration with NaOH:98.5 to 101.5 %
- Ethanol:≤0.5 %
Specification Test Results
Frequently Bought Together