Xanthohumol 97%
Supplier: Thermo Fisher Scientific
New Product
Certificates
About this item
CAS Number:6754-58-1
:
Formula:C₂₁H₂₂O₅
MDL Number:MFCD00210576
ADR:9,III
:354.40 g/mol
UN:3077
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:light yellow to dark yellow to orange powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Melting point:163°C to 173°C
Specification Test Results
Frequently Bought Together