
Dansyl hydrazide 97%
Catalog # 470230010.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:white to yellow to green crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Melting point:126°C to 130°C
Specification Test Results
About this item
:
:265.34 g/mol
Boiling Pt:450.8°C
EINECS:251-337-7
:GHS07
Storage Temperature:Ambient
CAS Number:33008-06-9
Formula:C₁₂H₁₅N₃O₂S
MDL Number:MFCD00003986