2,7-Dibromocarbazole
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 2,7-Dibromo-9H-carbazole
- CAS Number:136630-39-2
- Formula:C₁₂H₇Br₂N
- MDL Number:MFCD09033507
- Molecular Weight:325.00 g/mol
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
Synonyms: 2,7-Dibromo-9H-carbazole
- CAS Number:136630-39-2
- Formula:C₁₂H₇Br₂N
- MDL Number:MFCD09033507
- Molecular Weight:325.00 g/mol
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:white to yellow to beige powder and or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Melting point:230°C to 234°C
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



