Order Entry
Netherlands
ContactUsLinkComponent
3-Isochromanone
Irritant
3-Isochromanone
New Product
Certificates
Irritant
3-Isochromanone
New Product

Specification Test Results

  • Appearance:
    white to yellow crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Melting point:
    80°C to 84°C

Specification Test Results

Frequently Bought Together