Order Entry
Netherlands
ContactUsLinkComponent
Methanol, extra dry ≥99.9%, AcroSeal™
Toxic
Health
Flammable
Methanol, extra dry ≥99.9%, AcroSeal™
Catalog # 448411000.
CAS Number:  
undefined
Toxic
Health
Flammable
Methanol, extra dry ≥99.9%, AcroSeal™
Catalog # 448411000.
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 % (Coulometric)
  • GC:
    ≥99.85 %
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g
  • Filt.over 0.2 micron PFTE:
    Filtered

Specification Test Results

About this item

 :
 :32.04 g/mol
Boiling Pt:64.7 °C (760 mmHg)
Melting Pt:–98 °C
Density:0.791 g/cm³
Flash Pt:12 °C
UN:1230
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595