
N-Octyl-β-D-glucopyranoside
Catalog # DG016
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:292.37 g/mol
Melting Pt:110 °C
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
CAS Number:29836-26-8
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288