Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Florisil® for chromatography, Supelco®

Supplier:   Merck
Florisil® for chromatography, Supelco®
Supplier:  Merck
Certificates
Florisil® for chromatography, Supelco®
Florisil® for chromatography, Supelco®

2 Options Available Below

Product Details & Documents
Synonyms:  Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
  • CAS Number:1343-88-0
  • Formula:MgO₃Si
  • MDL Number:MFCD00078249
  • Molecular Weight:100.38 g/mol
  • EINECS:215-681-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 152,00 €
    152,00 €
    1 * 100 g
       152,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 607,00 €
    607,00 €
    1 * 500 g
       607,00 €