

Nitro blue tetrazolium chloride (NBT), yellow powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:298-83-9
:
Formula:C₄₀H₃₀Cl₂N₁₀O₆
MDL Number:MFCD00012159
:817.65 g/mol
Melting Pt:189…192 °C (lit.)
EINECS:206-067-4
:GHS08
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- identity Test:Passes
- l max (MeOH) :255 ± 3 nm
Specification Test Results
Frequently Bought Together