Membrane Filters, Nylon
Supplier: Advantech
New Product
Certificates
About this item
These membrane filters are inherently hydrophilic membranes made by impregnating a polyester web with nylon and are used for HPLC sample preparation, to sterilize biological solutions or buffers, or for vacuum degassing.
- High strength and heat resistance
- Binds proteins, DNA and RNA
- Pure, negligible organic extractables
- Maximum operating temperature: 180 °C
- Compatible with many aqueous and alcoholic solvents and solutions
Suitable for HPLC sample preparation. Filter sterilise and clarify aqueous and organic solvent solutions including buffers, microbiological and tissue culture solutions. Vacuum degassing.
Frequently Bought Together