Rhod-2 AM (Bromide) fluorescent indicator, for calcium determination
Catalog # STMC100-0994
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1124.0 g/mol
CAS Number:145037-81-6
Formula:C₅₂H₅₉BrN₄O₁₉