Precision Balance, L-Platform, XPR
Supplier: Mettler - Toledo
New Product
Certificates
About this item
Precision balance with exceptional results and smart quality assurance.
- Safe and easy cleaning
- Outstanding weighing performance
- Audit-Proof results every time
- Management of users and routine tests and consistent workflows
- StatusLight™ uses color to provide instant information on the balance status
The MonoBloc™ weighing cell delivers fast, accurate and reliable results, ensuring high productivity even in challenging weighing conditions. The StatusLight™, LevelControl, and GWP Approved provide at-a-glance information to verify your results are valid and ready for audit any time.
At-a-glance StatusLight™ Green means it is safe to weigh and results will be valid, yellow is a warning, perhaps that a routine test is due, and red means an issue must be resolved before weighing.
Specifications
- Catalog No:
- METB30316485
- METB30316701
- METB30316292
- METB30317428
- Communication interface:
- Bluetooth (optional), Ethernet (LAN), RS232 (integrated/optional), USB-A (to device), USB-B (to device)
- Bluetooth (optional), Ethernet (LAN), RS232 (integrated/optional), USB-A (to device), USB-B (to device)
- USB-B (to device), RS232 (integrated/optional), USB-A (to device), Bluetooth (optional), Ethernet (LAN)
- Bluetooth (optional), Ethernet (LAN), RS232 (integrated/optional), USB-A (to device), USB-B (to device)