Order Entry
Netherlands
ContactUsLinkComponent
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI5C5] (PE (Phycoerythrin))
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI5C5] (PE (Phycoerythrin))
Catalog # NOVUNBP2-70731PE
Supplier:  Novus Biologicals
New Product
Anti-FGFR2 Mouse Monoclonal Antibody [clone: OTI5C5] (PE (Phycoerythrin))
Catalog # NOVUNBP2-70731PE
Supplier:  Novus Biologicals
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibroblast growth factor receptor 2
  • Antigen symbol:
    FGFR2
  • Clonality:
    Monoclonal
  • Clone:
    OTI5C5
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    FGFR2
  • Antigen synonyms:
    TK14|CD332 antigen|ECT1|EC 2.7.10|TK25|BEK|soluble FGFR4 variant 4|K-sam|JWS|KGFR|CFD1|EC 2.7.10.1|craniofacial dysostosis 1|Keratinocyte growth factor receptorreceptor like 14|FLJ98662|CEK3|CD332|FGF R2|KSAM|BBDS|BFR-1|Jackson-Weiss syndrome
  • Purity:
    Immunogen affinity purified
  • Storage buffer:
    PBS, 0,05% sodium azide
  • Storage temperature:
    Store at 4 °C in the dark
  • Immunogen:
    Full length recombinant protein of human FGF R2 (NP_000132) produced in HEK293T cells
  • Pk:
    0,1 mL

Specifications

About this item

  • Dilutions: CyTOF-ready, flow cytometry, immunocytochemistry/immunofluorescence, immunohistochemistry, western blot
Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Type: Primary
Antigen: FGFR2
Clonality: Monoclonal
Clone: OTI5C5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, mouse, rat