Order Entry
Netherlands
ContactUsLinkComponent
Magicard™ Colour + Black Film for Magicard™ 300, 250 Shot
Magicard™ Colour + Black Film for Magicard™ 300, 250 Shot
Catalog # BRDY321972
Supplier:  Brady
New Product
undefined
Magicard™ Colour + Black Film for Magicard™ 300, 250 Shot
Catalog # BRDY321972
Supplier:  Brady
New Product

Specifications

  • Colour:
    Assorted
  • Description:
    250 shot colour film, 4-colour ribbon (Cyan, Magenta, Yellow, Black), 250 single-sided cards
  • Length:
    15 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Magicard™ 300

Specifications

About this item

The Magicard™ combined colour/black ribbon is perfect for full colour prints on one side and black prints on the other side, using a Magicard™ 300 printer.

  • High-quality, full colour ribbon for one side with overcoat and black on the other side
  • Includes a cleaning roller for dust free printing

Print 250 cards, full colour on one side with black on the reverse side.

Delivery: Includes 1 dye film and 1 cleaning roller.