Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Thiacloprid
Catalog # 14783-50MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Thiacloprid
Catalog # 14783-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • CAS No.:
    111988-49-9
  • Description:
    Thiacloprid
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H301 - H332 - H336 - H351 - H360FD - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD02101042
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    [3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidinylidene]cyanamide
  • Molecular weight:
    252,72

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P201 - P273 - P301 + P310 + P330 - P304 + P340 + P312