Supelco® Organic Reference Standard, Pullulan Standard
Catalog # 97873-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Certified reference material, certified according to DIN, for GPC, 12000
- Description:Pullulan Standard
- Seq:13
- Storage conditions:2…8 °C
- Shipping conditions:STD