Supelco® Organic Reference Standard, Maize GMO Standard for CHB-351 ‘Starlink’
Catalog # SUPL68993-1EA-F
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Description:Maize GMO Standard for CHB-351 ‘Starlink’
- Seq:10
- Storage conditions:2…8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):GMO Standard for Maize CHB-351 ‘Starlink’