Order Entry
Netherlands
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, 2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid
Supelco® TraceCERT® Organic Reference Standard, 2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid
Catalog # CRM79601-10MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, 2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid
Catalog # CRM79601-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    1450828-63-3
  • Description:
    2-Hydroxy-3-(1H-1,2,4-triazol-1-yl)propanoic acid
  • Storage conditions:
    2…8 °C
  • MDL No.:
    MFCD28515965
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    α-Hydroxy-1H-1,2,4-triazole-1-propanoic acid
  • Molecular weight:
    157,13

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.