Order Entry
Netherlands
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, 2-Amino-3-(1H-1,2,4-triazol-1-yl)propanoic acid
  CRM74418-10MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, 2-Amino-3-(1H-1,2,4-triazol-1-yl)propanoic acid
  CRM74418-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    114419-45-3
  • Description:
    2-Amino-3-(1H-1,2,4-triazol-1-yl)propanoic acid
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00077168
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    α-Amino-1H-1,2,4-triazole-1-propanoic acid; 3-(1,2,4-Triazol-1-yl)-ᴅʟ-alanine; H-β-(1,2,4-Triazol-1-yl)-ᴅʟ-Ala-OH
  • Molecular weight:
    156,14

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.