Supelco® TraceCERT® Organic Reference Standard, Maleic hydrazide
CRM67139-100MG
: Merck
:
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:123-33-1
- Description:Maleic hydrazide
- Storage conditions:15...25 °C
- Hazard class description:H315 - H319 - H335 - H341
- MDL No.:MFCD00006665
- M-Clarity quality level:MQ300
- Synonym(s):1,2-Dihydro-3,6-pyridazinedione; 3,6-Dihydroxypyridazine; 3,6-Pyridazinediol
- Melting point:299 - 301 °C (dec.) (lit.)
- Molecular weight:112,09
: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
: P202 - P261 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313