Order Entry
Netherlands
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Ethoprophos
Supelco® TraceCERT® Organic Reference Standard, Ethoprophos
Catalog # CRM53161-100MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Ethoprophos
Catalog # CRM53161-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    13194-48-4
  • Description:
    Ethoprophos
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H300 + H310 + H330 - H317 - H410
  • MDL No.:
    MFCD00055333
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    O-Ethyl S,S-dipropyl phosphorodithioate; Ethoprofos; Ethoprop
  • Molecular weight:
    242,34

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310