Order Entry
Netherlands
ContactUsLinkComponent
Supelco® Organic Reference Standard, Lutein (Zeaxanthin free)
Supelco® Organic Reference Standard, Lutein (Zeaxanthin free)
Catalog # 94110-1MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Lutein (Zeaxanthin free)
Catalog # 94110-1MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    Analytical standard
  • CAS No.:
    127-40-2
  • Description:
    Lutein (Zeaxanthin free)
  • Storage conditions:
    –20 °C
  • Shipping conditions:
    DICE
  • MDL No.:
    MFCD08435941
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Xanthophyll; α-Carotene-3,3′-diol; β, ε-carotene; β, ε-carotene-3,3′-diol; Lutein
  • Molecular weight:
    568,87

Specifications

About this item