Supelco® Organic Reference Standard, Suberic acid-2,3,4,5,6,7-¹³C₆
Supplier: Merck
Certificates
About this item
Caution: P264 - P280 - P305 + P351 + P338 - P337 + P313
Specifications
- Storage conditions:2…8 °C
- Hazard class description:H319
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Octanedioic acid-2,3,4,5,6,7-¹³C₆; Suberic acid-¹³C₆
- Molecular weight:180,15
Specifications
Frequently Bought Together