0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Netherlands
ContactUsLinkComponent
XFD750 amine
XFD750 amine
XFD750 amine
Catalog # AATB1710
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
XFD750 amine
Catalog # AATB1710
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
VPE:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
1155.17 g/mol
Product Details