Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Inloggen
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
Netherlands
Bestelling invoeren
Status van de bestelling
Uw account
Winkelwagen
ContactUsLinkComponent
Home
ATTO 647 maleimide
ATTO 647 maleimide
ATTO 647 maleimide
ATTO 647 maleimide
Catalog # AATB2853
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
ATTO 647 maleimide
Catalog # AATB2853
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
828.95 g/mol
Product Details