Anti-USP44 Rabbit Polyclonal Antibody
ANTIA100725-100
New Product
- Antibody type:Primary
- Antigen name:Deubiquitinating enzyme 44
- Antigen symbol:USP44
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9H0E7
- Antigen synonyms:ubiquitin specific peptidase 44|E430004F17Rik|Ubiquitin carboxyl-terminal hydrolase 44|Ubiquitin-specific-processing protease 44|UBP44_HUMAN|Ubiquitin thiolesterase 44|USP44
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:81 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human USP44 (amino acids 211 - 260)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to USP44 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: USP44
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human