Anti-CD5 Mouse Monoclonal Antibody [clone: ABT-CD5]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (ABT-CD5) antibody to CD5 for IHC with samples derived from human.
- Validated Applications: IHC
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: ABT-CD5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA100703-50
- ANTIA100703-100
- Antigen symbol:
- CD5
- CD5
- Antigen name:
- CD 5
- CD 5
- Antigen synonyms:
- CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
- CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-CD5
- ABT-CD5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P06127
- UniprotID# P06127
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human CD5
- Synthetic peptide derived from human CD5
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
- IHC
Specifications
Frequently Bought Together