Order Entry
Netherlands
ContactUsLinkComponent
Anti-Fibrillarin Mouse Monoclonal Antibody [clone: 38F3]
Anti-Fibrillarin Mouse Monoclonal Antibody [clone: 38F3]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-Fibrillarin Mouse Monoclonal Antibody [clone: 38F3]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA85370-250
  • ANTIA85370-500
  • Antigen symbol:
  • Fibrillarin
  • Fibrillarin
  • Antigen name:
  • 34 kD nucleolar scleroderma antigen
  • 34 kD nucleolar scleroderma antigen
  • Antigen synonyms:
  • FIB|FIB1|Fibrillarin|FBRL_HUMAN|Fbl|Histone-glutamine methyltransferase|FLRN|34 kDa nucleolar scleroderma antigen|Nop1p
  • FIB|FIB1|Fibrillarin|FBRL_HUMAN|Fbl|Histone-glutamine methyltransferase|FLRN|34 kDa nucleolar scleroderma antigen|Nop1p
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 38F3
  • 38F3
  • Reactivity:
  • Zebrafish, Yeast, S. cerevisiae, Drosophila, C. elegans, Human, Rat, Mouse
  • Zebrafish, Yeast, S. cerevisiae, Drosophila, C. elegans, Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P15646
  • UniprotID# P15646
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Yeast nuclear preparation
  • Yeast nuclear preparation
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 35 kDa
  • 35 kDa
  • Storage buffer:
  • Supplied as an aliquot of concentrated hybridoma cell culture media with 5 mM sodium azide
  • Supplied as an aliquot of concentrated hybridoma cell culture media with 5 mM sodium azide
  • Storage temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • ICC/IF, IHC
  • ICC/IF, IHC

Specifications

Frequently Bought Together