Anti-PI 3 Kinase p85 beta Rabbit Polyclonal Antibody
ANTIA94496-100
New Product
- Antibody type:Primary
- Antigen name:p85
- Antigen symbol:PI 3 Kinase p85 beta
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O00459
- Antigen synonyms:Phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 2 (p85 beta)|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 2|Phosphatidylinositol 3 kinase regulatory beta subunit|Phosphatidylinositol 3 kinase|P85B_HUMAN|Phosphatidylinositol 3 kinase regulatory subunit beta|Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta|p85 beta|P85B
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:81 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PI 3-kinase p85beta (amino acids 409-458)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to PI 3-kinase p85beta for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: PI 3 Kinase p85 beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat