Order Entry
Netherlands
ContactUsLinkComponent
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [Clone: C-43] (FITC (Fluorescein Isothiocyanate))
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [Clone: C-43] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86583-100
Supplier:  Antibodies.com
New Product
Anti-Cytokeratin 8 Mouse Monoclonal Antibody [Clone: C-43] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86583-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CARD2
  • Antigen symbol:
    Cytokeratin 8
  • Clonality:
    Monoclonal
  • Clone:
    C-43
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Rabbit,
    Sheep,
    Human,
    Bovine
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q7L4M3
  • Antigen synonyms:
    Cytokeratin endo A|CYK8|CK-8|CK 8|DreK8|Cytokeratin-8|CYKER|CK8|Cytokeratin 8
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Cytoskeleton preparation from HeLa human cervix carcinoma cell line
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [C-43] antibody to Cytokeratin 8 (FITC) for Flow Cytometry with samples derived from Human, Sheep, Bovine, Rabbit and Porcine.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 8
Clonality: Monoclonal
Clone: C-43
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Sheep, Bovine, Rabbit, Porcine