Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-HLA E Mouse Monoclonal Antibody [Clone: MEM-E/02]
  ANTIA86105-100
New Product
Anti-HLA E Mouse Monoclonal Antibody [Clone: MEM-E/02]
  ANTIA86105-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    EA1.2
  • Antigen symbol:
    HLA E
  • Clonality:
    Monoclonal
  • Clone:
    MEM-E/02
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13747
  • Antigen synonyms:
    EA2.1|HLA class I histocompatibility antigen E alpha chain precursor|HLA E|HLA 6.2|HLA6.2|Lymphocyte antigen|HLAE|HLA class I histocompatibility antigen alpha chain E|HLA class I histocompatibility antigen E alpha chain
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human HLA-E denaturated heavy chain
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [MEM-E/02] antibody to HLA E for WB and IHC-P with samples derived from Human and Non-Human Primates.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: IHC-P: 5 to 10 µg/ml

Type: Primary
Antigen: HLA E
Clonality: Monoclonal
Clone: MEM-E/02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates