Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD9 Mouse Monoclonal Antibody [Clone: MEM-61] (APC (Allophycocyanin))
Anti-CD9 Mouse Monoclonal Antibody [Clone: MEM-61] (APC (Allophycocyanin))
Catalog # ANTIA86086-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD9 Mouse Monoclonal Antibody [Clone: MEM-61] (APC (Allophycocyanin))
Catalog # ANTIA86086-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5H9
  • Antigen symbol:
    CD9
  • Clonality:
    Monoclonal
  • Clone:
    MEM-61
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P21926
  • Antigen synonyms:
    Antigen defined by monoclonal|BTCC1|CD9 antigen p24|BA-2/p24 antigen|BTCC 1|BA2|CD9 antigen|5H9 antigen|CD9
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Pre-B cell line NALM-6
  • Purification:
    This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-61] antibody to CD9 (APC) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: MEM-61
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human