Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD48 Mouse Monoclonal Antibody [Clone: MEM-102] (PE (Phycoerythrin))
Anti-CD48 Mouse Monoclonal Antibody [Clone: MEM-102] (PE (Phycoerythrin))
Catalog # ANTIA86027-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD48 Mouse Monoclonal Antibody [Clone: MEM-102] (PE (Phycoerythrin))
Catalog # ANTIA86027-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen CD48
  • Antigen symbol:
    CD48
  • Clonality:
    Monoclonal
  • Clone:
    MEM-102
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P09326
  • Antigen synonyms:
    BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Raji human Burkitt's lymphoma cell line
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-102] antibody to CD48 (PE) for Flow Cytometry with samples derived from Human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: MEM-102
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates