Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CD74 Rabbit Recombinant Antibody [clone: CLIP/7023R]
  ANTIA278102-100
New Product
undefined
Anti-CD74 Rabbit Recombinant Antibody [clone: CLIP/7023R]
  ANTIA278102-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD 74
  • Antigen symbol:
    CD74
  • Clonality:
    Recombinant
  • Clone:
    CLIP/7023R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Baboon,
    Human,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P044233
  • Antigen synonyms:
    HG2A_HUMAN|Gamma chain of class II antigens|CD74 antigen|CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated)|CLIP|CD74 molecule|CD74 molecule, major histocompatibility complex, class II invariant chain|CD74|DHLAG
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    33 - 41 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    SU-DHL-4 lymphoma cells.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [CLIP/7023R] antibody to CD74 for IHC-P with samples derived from human, babbon and mouse.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD74
Clonality: Recombinant
Clone: CLIP/7023R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Baboon;Mouse