Anti-Cadherin 16 Rabbit Recombinant Antibody [clone: CDH16/7027R]
Catalog # ANTIA277948-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CAD16_HUMAN
- Antigen symbol:Cadherin 16
- Clonality:Recombinant
- Clone:CDH16/7027R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# O75309
- Antigen synonyms:CDH16|Cadherin-16|CDH 16|Kidney specific cadherin|Cadherin 16 KSP cadherin|Cadherin 16|Kidney cadherin|Cadherin kidney|Kidney-specific cadherin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:130 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Cadherin 16.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CDH16/7027R] antibody to Cadherin 16 for IHC-P with samples derived from human, mouse and rat.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cadherin 16
Clonality: Recombinant
Clone: CDH16/7027R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat