Anti-CD99 Mouse Monoclonal Antibody [clone: SPM586]
ANTIA254328-100
New Product
- Antibody type:Primary
- Antigen name:120000000
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:SPM586
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human,Rat
- Form:Liquid
- Gene ID:UniprotID# P14209
- Antigen synonyms:CD99|CD99_HUMAN|Antigen identified by monoclonal antibodies 12E7, F21 and O13|CD99 molecule|Cell surface antigen O13|Antigen identified by monoclonal 12E7, Y homolog|Cell surface antigen 12E7|CD99 antigen|Cell surface antigen HBA 71
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified E-rosette forming cells from human peripheral blood lymphocytes.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [SPM586] antibody to CD99 for Flow Cytometry, IF and IHC-P with samples derived from human and rat.
- Validated Applications: Flow Cytometry, IF, IHC-P
- Recommended Dilutions: Flow Cytometry: 5 to 10 µl / million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD99
Clonality: Monoclonal
Clone: SPM586
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human;Rat