Order Entry
Netherlands
ContactUsLinkComponent
Anti-CD5 Rabbit Recombinant Antibody [Clone: C5/6463R]
Anti-CD5 Rabbit Recombinant Antibody [Clone: C5/6463R]
Catalog # ANTIA278091-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD5 Rabbit Recombinant Antibody [Clone: C5/6463R]
Catalog # ANTIA278091-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 5
  • Antigen symbol:
    CD5
  • Clonality:
    Recombinant
  • Clone:
    C5/6463R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06127
  • Antigen synonyms:
    CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    67 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CD5. The immunogen contains the specific extracellular domain of human CD5 (D32-P372).
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [C5/6463R] antibody to CD5 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD5
Clonality: Recombinant
Clone: C5/6463R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human