Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-HSP27 Rabbit Recombinant Antibody [Clone: HSPB1/7038R]
  ANTIA277997-100
New Product
undefined
Anti-HSP27 Rabbit Recombinant Antibody [Clone: HSPB1/7038R]
  ANTIA277997-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    28 kDa heat shock protein
  • Antigen symbol:
    HSP27
  • Clonality:
    Recombinant
  • Clone:
    HSPB1/7038R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04792
  • Antigen synonyms:
    epididymis secretory protein Li 102|Heat shock 27 kDa protein|Estrogen regulated 24 kDa protein|Heat shock 27kD protein 1|Heat shock 27kDa protein|Estrogen-regulated 24 kDa protein|DKFZp586P1322|CMT2F|Heat shock 25kDa protein 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    27 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human HSP27 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal [HSPB1/7038R] antibody to HSP27 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HSP27
Clonality: Recombinant
Clone: HSPB1/7038R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human