Anti-Keratin 76 Rabbit Recombinant Antibody [Clone: KRTH/1576R]
ANTIA254190-100
New Product
- Antibody type:Primary
- Antigen symbol:Keratin 76
- Clonality:Recombinant
- Clone:KRTH/1576R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q01546
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:52 - 67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KRT76 protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [KRTH/1576R] antibody to Keratin 76 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: Keratin 76
Clonality: Recombinant
Clone: KRTH/1576R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human