Anti-Keratin 76 Mouse Recombinant Antibody [Clone: rKRTH/2148]
ANTIA254134-100
New Product
- Antibody type:Primary
- Antigen symbol:Keratin 76
- Clonality:Recombinant
- Clone:rKRTH/2148
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q01546
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:50 - 67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Solubilized keratin extract from human stratum corneum.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rKRTH/2148] antibody to Keratin 76 for IHC-P with samples derived from human, mouse and rat.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Keratin 76
Clonality: Recombinant
Clone: rKRTH/2148
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat