Anti-GRP94 Rabbit Recombinant Antibody [clone: HSP90B1/3168R]
ANTIA253388-100
New Product
- Antibody type:Primary
- Antigen name:94 kDa glucose regulated protein
- Antigen symbol:GRP94
- Clonality:Recombinant
- Clone:HSP90B1/3168R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Dog,Rat,Mouse,Hamster,Pig,Rabbit,Horse,Monkey,Sheep,Chicken,Human,Bovine,Guinea pig,Xenopus laevis
- Form:Liquid
- Gene ID:UniprotID# P14625
- Antigen synonyms:ECGP|GRP 94|Endothelial cell (HBMEC) glycoprotein|ENPL_HUMAN|Endoplasmin|94 kDa glucose-regulated protein|gp96|Glucose regulated protein 94kDa|gp96 homolog
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:94 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human HSP90B1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [HSP90B1/3168R] antibody to GRP94 for Flow Cytometry and IHC-P with samples derived from human,monkey, horse, bovine, sheep, pig, rabbit, canine, rat, mouse, hamster, guinea pig, chicken and xenopus laevis.
- Validated Applications: Flow Cytometry, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: GRP94
Clonality: Recombinant
Clone: HSP90B1/3168R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Horse, Bovine, Sheep, Pig, Rabbit, Dog, Rat, Mouse, Hamster, Guinea Pig, Chicken, Xenopus laevis