Order Entry
Netherlands
ContactUsLinkComponent
Anti-p53 Mouse Recombinant Antibody [clone: rBP53-12]
Anti-p53 Mouse Recombinant Antibody [clone: rBP53-12]
Catalog # ANTIA253356-100
Supplier:  Antibodies.com
New Product
undefined
Anti-p53 Mouse Recombinant Antibody [clone: rBP53-12]
Catalog # ANTIA253356-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen NY-CO-13
  • Antigen symbol:
    p53
  • Clonality:
    Recombinant
  • Clone:
    rBP53-12
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Hamster,
    Monkey,
    Chicken,
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04637
  • Antigen synonyms:
    P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    53 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human wild type p53 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rBP53-12] antibody to p53 for WB and IHC-P with samples derived from human,monkey, canine, hamster and chicken.

  • Validated Applications: WB, IHC-P
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: p53
Clonality: Recombinant
Clone: rBP53-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Dog, Hamster, Chicken