Order Entry
Netherlands
ContactUsLinkComponent
Anti-Integrin alpha 4 Rabbit Polyclonal Antibody
Anti-Integrin alpha 4 Rabbit Polyclonal Antibody
Catalog # ANTIA95690-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Integrin alpha 4 Rabbit Polyclonal Antibody
Catalog # ANTIA95690-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    269C wild type
  • Antigen symbol:
    Integrin alpha 4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13612
  • Antigen synonyms:
    CD49d|Antigen CD49D, alpha 4 subunit of VLA 4 receptor|CD49 antigen-like family member D|CD49 antigen like family member D|Integrin alpha IV|IA4|Integrin alpha-4|Integrin alpha 4 subunit|Integrin alpha 4
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    115 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Integrin alpha4 (amino acids 974-1023)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to Integrin alpha4 for WB and ELISA with samples derived from Human and Mouse.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Integrin alpha 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse